α-Bromo-2,3,4,5-tetrafluorotoluene ≥97%

Supplier: Matrix Scientific

Synonyms: 2,3,4,5-Tetrafluorobenzyl bromide

003457-1G 003457-5G
101776-730EA 53.85 USD
101776-730 101776-732
α-Bromo-2,3,4,5-tetrafluorotoluene ≥97%
α-Bromo-2,3,4,5-tetrafluorotoluene

Hazard: Corrosive/Lachrymator

Formula: C₇H₃BrF₄
Boiling Pt: 107-110°C/65mm
MDL Number: MFCD01631453
CAS Number: 53001-71-1
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 107-110°/6
ND20 1.487


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR