2,3,4,5-Tetrafluorobenzoyl chloride ≥95%

Supplier: Matrix Scientific

Synonyms: 2,3,4,5-tetrafluorobenzoylchloride

003441-25G 003441-100G
101776-724EA 53.85 USD
101776-724 101776-726
2,3,4,5-Tetrafluorobenzoyl chloride ≥95%
2,3,4,5-Tetrafluorobenzoyl chloride

Hazard: Corrosive/Moisture Sensitive/Lachrymator

Formula: C₇HClF₄O
MW: 212.53 g/mol
Density: 1.58
MDL Number: MFCD00075164
CAS Number: 94695-48-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Density 1.58
ND20 1.4787

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR