1-Ethyl-2-iodobenzene ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Ethyliodobenzene, 1-Iodo-2-ethylbenzene

003407-25G 003407-100G
101776-712EA 30.77 USD
101776-712 101776-714
1-Ethyl-2-iodobenzene ≥97%
1-Ethyl-2-iodobenzene

Hazard: Irritant

Formula: C₈H₉I
MW: 232.06 g/mol
Boiling Pt: 226 °C
Density: 1.618
MDL Number: MFCD00040871
CAS Number: 18282-40-1

Order Now

Specification Test Results

Boiling point 226°
ND20 1.5959


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR