Ethyl-2-methyl-4,4,4-trifluoroacetoacetate, Technical Grade

Supplier: Matrix Scientific

Synonyms: Ethyl 4,4,4-Trifluoro-2-methyl-3-oxobutyrate

003398-5G 003398-25G
101776-704EA 87.69 USD
101776-704 101776-706
Ethyl-2-methyl-4,4,4-trifluoroacetoacetate, Technical Grade
Ethyl-2-methyl-4,4,4-trifluoroacetoacetate

Hazard: Flammable

Formula: C₇H₉F₃O₃
MW: 198.14 g/mol
Boiling Pt: 144…146 °C
Density: 1.258
MDL Number: MFCD00190635
CAS Number: 344-00-3

Order Now

Specification Test Results

Boiling point 144-146°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR