Ethyl-2-methyl-4,4,4-trifluorobutyrate ≥95%

Supplier: Matrix Scientific

003397-25G 003397-1G
101776-702EA 795.38 USD
101776-702 101776-698
Ethyl-2-methyl-4,4,4-trifluorobutyrate ≥95%
Ethyl-2-methyl-4,4,4-trifluorobutyrate

Hazard: Flammable

Formula: C₇H₁₁F₃O₂
MW: 184.16 g/mol
Boiling Pt: 142-145°C
MDL Number: MFCD00190636
CAS Number: 143484-00-8

Order Now

Specification Test Results

Boiling point 142-145°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR