Ethyl-3-methyl-4,4,4-trifluorobutyrate ≥97%

Supplier: Matrix Scientific

003396-1G 003396-5G
101776-694EA 36.92 USD
101776-694 101776-696
Ethyl-3-methyl-4,4,4-trifluorobutyrate ≥97%
Ethyl-3-methyl-4,4,4-trifluorobutyrate

Hazard: Flammable

Formula: C₇H₁₁F₃O₂
MW: 184.16 g/mol
Boiling Pt: 135…136 °C
MDL Number: MFCD00040847
CAS Number: 6975-13-9

Order Now

Specification Test Results

Boiling point 135-136°
ND20 1.3602

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR