3-Fluorobenzylamine ≥98%

Supplier: Matrix Scientific

003350-25G 003350-100G 003350-5G
101776-684EA 223.08 USD
101776-684 101776-686 101776-682
3-Fluorobenzylamine ≥98%
3-Fluorobenzylamine

Hazard: Corrosive

Formula: C₇H₈FN
Boiling Pt: 87…89 °C (25 mmHg)
Density: 1.097
MDL Number: MFCD00008113
CAS Number: 100-82-3
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Boiling point 87-89°/25m
Density 1.097
ND20 1.514


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR