3-Chlorobenzyl isothiocyanate ≥98%

Supplier: Matrix Scientific

003340-5G 003340-1G
101776-664EA 489.23 USD
101776-664 101776-662
3-Chlorobenzyl isothiocyanate ≥98%
3-Chlorobenzyl isothiocyanate

Hazard: Toxic/Moisture Sensitive/Lachrymator

Formula: C₈H₆ClNS
MW: 183.66
Boiling Pt: 140 °C (6 mmHg)
MDL Number: MFCD00046820
CAS Number: 3694-58-4

Order Now

Specification Test Results

Boiling point 140°/6mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR