4-Fluorobenzyl isothiocyanate ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Fluorobenzylisothiocyanate, 1-Fluoro-4-(isothiocyanatomethyl)benzene
003336-1G
003336-5G
101776-650EA
21.54
USD
101776-650
101776-652
4-Fluorobenzyl isothiocyanate ≥97%
4-Fluorobenzyl isothiocyanate
Hazard: Toxic/Refrigerate/Lachrymator
Formula:
C₈H₆FNS MW: 167.21 g/mol Boiling Pt: 125°C/6mm Density: 1.22 |
MDL Number:
MFCD00041115 CAS Number: 2740-88-7 |
Specification Test Results
Boiling point | 125°/6mm |
Density | 1.22 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...