α-Bromo-2-fluorotoulene ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Fluorobenzyl bromide, α-Bromo-o-fluorotoulene

003328-100G 003328-25G
101776-644EA 69.23 USD
101776-644 101776-642
α-Bromo-2-fluorotoulene ≥98%
α-Bromo-2-fluorotoulene

Hazard: Corrosive/Lachrymator

Formula: C₇H₆BrF
Boiling Pt: 84°C/15mm
Density: 1.567
MDL Number: MFCD00000324
CAS Number: 446-48-0
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 84°/15mm
Density 1.567
ND20 1.552


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR