α-Bromo-2-chlorotoulene ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(Bromomethyl)-2-chlorobenzene, 2-Chlorobenzyl bromide

003327-100G 003327-25G
101776-640EA 420 USD
101776-640 101776-638
α-Bromo-2-chlorotoulene ≥97%
α-Bromo-2-chlorotoulene

Hazard: Corrosive/Refrigerate/Lachrymator

Formula: C₇H₆BrCl
MW: 205.48 g/mol
Boiling Pt: 103-104°C/10mm
Density: 1.583
MDL Number: MFCD00000566
CAS Number: 611-17-6
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 103-104°/1
Density 1.583
ND20 1.592

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR