2-Chlorobenzoyl isothiocyanate ≥97%

Supplier: Matrix Scientific

003320-5G 003320-1G
101776-636EA 258.46 USD
101776-636 101776-634
2-Chlorobenzoyl isothiocyanate ≥97%
2-Chlorobenzoyl isothiocyanate

Hazard: Corrosive/Irritant/Store in Freezer

Formula: C₈H₄ClNOS
Boiling Pt: 130 °C (5 mmHg)
MDL Number: MFCD00037829
CAS Number: 5067-90-3

Order Now

Specification Test Results

Boiling point 130°/5mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR