3-Chlorobenzoyl isothiocyanate ≥97%

Supplier: Matrix Scientific

003318-5G 003318-1G
101776-628EA 176.92 USD
101776-628 101776-626
3-Chlorobenzoyl isothiocyanate ≥97%
3-Chlorobenzoyl isothiocyanate

Hazard: Corrosive/Irritant/Store in Freezer

Formula: C₈H₄ClNOS
MDL Number: MFCD00041103
CAS Number: 66090-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR