4-Fluorobenzoyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: p-Fluorobenzoyl chloride

003316-100G 003316-500G
101776-622EA 146.15 USD
101776-622 101776-624
4-Fluorobenzoyl chloride ≥98%
4-Fluorobenzoyl chloride

TCA listed: T
Hazard: Corrosive/Lachrymator

Formula: C₇H₄ClFO
Boiling Pt: 82°C/20mm
Melting Pt: 9°C
Density: 1.342
MDL Number: MFCD00000684
CAS Number: 403-43-0
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 82°/20mm
Density 1.342
ND20 1.5309


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR