3-Fluorobenzoyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Fluorobenzoylchloride, m-Fluorobenzoyl chloride

003315-100G 003315-25G
101776-620EA 275.38 USD
101776-620 101776-618
3-Fluorobenzoyl chloride ≥98%
3-Fluorobenzoyl chloride

TCA listed: T
Hazard: Corrosive/Lachrymator

Formula: C₇H₄ClFO
MW: 158.56 g/mol
Boiling Pt: 189 °C
Melting Pt: –30 °C
Density: 1.304
MDL Number: MFCD00000670
CAS Number: 1711-07-5
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 189°
Density 1.304
ND20 1.5285

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR