α,α,α,2-Tetrafluorotoluene ≥99%
Supplier: Matrix Scientific
Synonyms:
1-Fluoro-2-(trifluoromethyl)benzene, 2-Fluorobenzotrifluoride
003308-100G
003308-500G
101776-602EA
35.38
USD
101776-602
101776-604
α,α,α,2-Tetrafluorotoluene ≥99%
α,α,α,2-Tetrafluorotoluene
TCA listed: T
Hazard: Flammable
Formula:
C₇H₄F₄ MW: 164.1 g/mol Boiling Pt: 114-115°C/750mm Density: 1.293 |
MDL Number:
MFCD00000374 CAS Number: 392-85-8 UN: 1993 ADR: 3,II |
Specification Test Results
Boiling point | 114-115°/7 |
Density | 1.293 |
ND20 | 1.406 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...