α,α,α,4-Tetrafluorotoluene ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Fluorobenzotrifluoride, 1-Fluoro-4-(trifluoromethyl)benzene

003307-100G 003307-25G
101776-600EA 55.38 USD
101776-600 101776-598
α,α,α,4-Tetrafluorotoluene ≥98%
α,α,α,4-Tetrafluorotoluene

TCA listed: T
Hazard: Flammable

Formula: C₇H₄F₄
MW: 164.1 g/mol
Boiling Pt: 102-105°C
Melting Pt: -42 to -41.7°C
Density: 1.293
MDL Number: MFCD00000399
CAS Number: 402-44-8
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Boiling point 102-105°
Density 1.293
ND20 1.401

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR