α,α,α-Trifluoro-o-xylene ≥97%

Supplier: Matrix Scientific

003305-1G 003305-5G
101776-590EA 13.85 USD
101776-590 101776-592
α,α,α-Trifluoro-o-xylene ≥97%
α,α,α-Trifluoro-o-xylene

Hazard: Flammable

Formula: C₈H₇F₃
Boiling Pt: 125…126 °C
MDL Number: MFCD00153204
CAS Number: 13630-19-8

Order Now

Specification Test Results

Boiling point 125-126°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR