2-Chlorobenzenesulphonyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Chlorobenzene-1-sulphonyl chloride

003274-25G 003274-5G
101776-588EA 58.46 USD
101776-588 101776-586
2-Chlorobenzenesulphonyl chloride ≥98%
2-Chlorobenzenesulphonyl chloride

TCA listed: T
Hazard: Corrosive/Moisture Sensitive

Formula: C₆H₄Cl₂O₂S
Boiling Pt: 150…152 °C (12 mmHg)
Melting Pt: 26…28 °C
MDL Number: MFCD00051520
CAS Number: 2905-23-9
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 150-152°/1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR