2-Fluorobenzenesulphonyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Fluorobenzene-1-sulphonyl chloride

003273-25G 003273-100G
101776-582EA 29.23 USD
101776-582 101776-584
2-Fluorobenzenesulphonyl chloride ≥98%
2-Fluorobenzenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive/Refrigerate

Formula: C₆H₄ClFO₂S
MW: 194.61
Boiling Pt: 242
Melting Pt: 26 °C
Density: 1.47
MDL Number: MFCD00042285
CAS Number: 2905-21-7
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Boiling point 143-146°/1
Density 1.47
ND20 1.537


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR