3-Fluorobenzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Fluorobenzene-1-sulphonyl chloride

003272-5G 003272-25G 003272-100G
101776-576EA 20 USD
101776-576 101776-578 101776-580
3-Fluorobenzenesulfonyl chloride ≥97%
3-Fluorobenzenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₆H₄ClFO₂S
MW: 194.61 g/mol
Boiling Pt: 110…112 °C (15 mmHg)
MDL Number: MFCD00042286
CAS Number: 701-27-9
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 110-112°/1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR