2-Fluorobenzene-1,3-diacetonitrile ≥97%

Supplier: Matrix Scientific

003267-250MG 003267-1G
101776-568EA 40 USD
101776-568 101776-570
2-Fluorobenzene-1,3-diacetonitrile ≥97%
2-Fluorobenzene-1,3-diacetonitrile

Hazard: Toxic

Formula: C₁₀H₇FN₂
Melting Pt: 75…77 °C
MDL Number: MFCD00052861
CAS Number: 175136-84-2

Order Now

Specification Test Results

Melting point 75-77°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR