3-Fluorobenzal chloride ≥98%

Supplier: Matrix Scientific

Synonyms: α,α-Dichloro-3-fluortoluene, 1-(Dichloromethyl)-3-fluorobenzene

003251-5G 003251-25G
101776-560EA 110.77 USD
101776-560 101776-562
3-Fluorobenzal chloride ≥98%
3-Fluorobenzal chloride

Hazard: Corrosive/Lachrymator

Formula: C₇H₅Cl₂F
MW: 179.02 g/mol
Boiling Pt: 195…197 °C
MDL Number: MFCD00037112
CAS Number: 402-64-2
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 195-197°
ND20 1.5275

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR