4-Nitrophenylsulfur pentafluoride ≥97%
Supplier: Matrix Scientific
003248-5G
003248-1G
101776-558EA
193.85
USD
101776-558
101776-556
4-Nitrophenylsulfur pentafluoride ≥97%
4-Nitrophenylsulphur pentafluoride
Hazard: Toxic/Irritant
Formula:
C₆H₄F₅NO₂S Melting Pt: 36…38 °C |
MDL Number:
MFCD00221619 CAS Number: 2613-27-6 UN: 2811 ADR: 6.1,III |
Specification Test Results
Melting point | 36-38° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...