1-Bromo-4-(trifluoromethylthio)benzene ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(Trifluoromethylthio)bromobenzene, 4-Bromophenyl trifluoromethyl sulphide

003232-5G 003232-25G
101776-552EA 113.85 USD
101776-552 101776-554
1-Bromo-4-(trifluoromethylthio)benzene ≥97%
1-Bromo-4-(trifluoromethylthio)benzene

Hazard: Irritant/Stench

Formula: C₇H₄BrF₃S
MW: 257.07 g/mol
Boiling Pt: 81°C/15mm
MDL Number: MFCD00040840
CAS Number: 333-47-1

Order Now

Specification Test Results

Boiling point 81°/15mm
ND20 1.5172

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR