4-[(Trifluoromethyl)thio]aniline ≥98%

Supplier: Matrix Scientific

Synonyms: 4-((Trifluoromethyl)thio)aniline

003230-100G 003230-5G 003230-25G
101776-548EA 446.15 USD
101776-548 101776-544 101776-546
4-[(Trifluoromethyl)thio]aniline ≥98%
4-[(Trifluoromethyl)thio]aniline

Hazard: Toxic/Stench

Formula: C₇H₆F₃NS
MW: 193.19 g/mol
Boiling Pt: 103 °C (11 mmHg)
Density: 1.351
MDL Number: MFCD00040926
CAS Number: 372-16-7
UN: 2810
ADR: 6.1 III

Order Now

Specification Test Results

Boiling point 103°/11mm
Density 1.351
ND20 1.528


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR