(±)-3-Methylvaleric acid ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Methylvaleric Acid, 3-Methylpentanoic acid, (±)-3-Methylpentanoic acid
003195-25G
003195-100G
101776-524EA
193.85
USD
101776-524
101776-526
(±)-3-Methylvaleric acid ≥97%
(±)-3-Methylvaleric acid
TCA listed: T
Hazard: Corrosive/Stench
Formula:
C₆H₁₂O₂ MW: 116.16 g/mol Boiling Pt: 196…198 °C Density: 0.93 |
MDL Number:
MFCD00002729 CAS Number: 105-43-1 UN: 3265 ADR: 8,III |
Specification Test Results
Boiling point | 196-198° |
Density | 0.93 |
ND20 | 1.4159 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...