1,1-Cyclobutanedicarboxylic acid ≥98%
Supplier: Matrix Scientific
003049-100G
003049-25G
101776-482EA
135.38
USD
101776-482
101776-480
1,1-Cyclobutanedicarboxylic acid ≥98%
1,1-Cyclobutanedicarboxylic acid
TCA listed: T
Hazard: Corrosive
Formula:
C₆H₈O₄ Melting Pt: 158 °C |
MDL Number:
MFCD00001325 CAS Number: 5445-51-2 UN: 3261 ADR: 8,III |
Specification Test Results
Melting point | 158° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...