2,3,5-Tribromothiophene ≥98%
Supplier: Matrix Scientific
003047-100G
003047-25G
101776-478EA
95.38
USD
101776-478
101776-476
2,3,5-Tribromothiophene ≥98%
2,3,5-Tribromothiophene
TCA listed: T
Hazard: Toxic
Formula:
C₄HBr₃S MW: 320.83 g/mol Boiling Pt: 120 °C (11 mmHg) Melting Pt: 28…29 °C Density: 2.483 |
MDL Number:
MFCD00014521 CAS Number: 3141-24-0 UN: 2811 ADR: 6.1,III |
Specification Test Results
Boiling point | 120°/11mm |
Density | 2.483 |
ND20 | 1.664 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...