3-Phenoxybenzyl chloride ≥98%
Supplier: Matrix Scientific
003027-1G
003027-5G
003027-25G
101776-470EA
49.23
USD
101776-470
101776-472
101776-474
3-Phenoxybenzyl chloride ≥98%
3-Phenoxybenzyl chloride
TCA listed: T
Hazard: Corrosive/Lachrymator
Formula:
C₁₃H₁₁ClO MW: 218.68 g/mol Boiling Pt: 142°C/5mm Density: 1.16 |
MDL Number:
MFCD00040866 CAS Number: 53874-66-1 UN: 3265 ADR: 8,III |
Specification Test Results
Boiling point | 142°/5mm |
Density | 1.16 |
ND20 | 1.591 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...