2-Chloropyrazine ≥98%

Supplier: Matrix Scientific

Synonyms: Chloropyrazine

002998-25G 002998-100G
101776-460EA 21.54 USD
101776-460 101776-462
2-Chloropyrazine ≥98%
2-Chloropyrazine

TCA listed: T
Hazard: Flammable/Stench

Formula: C₄H₃ClN₂
MW: 114.53 g/mol
Boiling Pt: 153-154°C
Density: 1.283
MDL Number: MFCD00006124
CAS Number: 14508-49-7
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 153-154°
Density 1.283
ND20 1.532

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR