4-(Diethylamino)phenyl isothiocyanate ≥98%

Supplier: Matrix Scientific

002972-1G 002972-5G
101776-456EA 30.77 USD
101776-456 101776-458
4-(Diethylamino)phenyl isothiocyanate ≥98%
4-(Diethylamino)phenyl isothiocyanate

Hazard: Toxic/Corrosive/Refrigerate/Lachrymator

Formula: C₁₁H₁₄N₂S
Boiling Pt: 147-148°C/1.2mm
Melting Pt: 27-29°C
MDL Number: MFCD00041094
CAS Number: 84381-54-4

Order Now

Specification Test Results

Boiling point 147-148°/1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR