11H-Eicosafluoroundecanoyl chloride ≥92%

Supplier: Matrix Scientific

002956-5G 002956-25G
101776-452EA 430.77 USD
101776-452 101776-454
11H-Eicosafluoroundecanoyl chloride ≥92%
11H-Eicosafluoroundecanoyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₁₁HClF₂₀O
Melting Pt: 54…56 °C
MDL Number: MFCD00042119
CAS Number: 2248-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR