Allylpentafluorobenzene ≥97%
Supplier: Matrix Scientific
Synonyms:
3-(Pentafluorophenyl)-1-propene, 1-Allyl-2,3,4,5,6-pentafluorobenzene
002945-5G
002945-25G
101776-448EA
29.23
USD
101776-448
101776-450
Allylpentafluorobenzene ≥97%
Allylpentafluorobenzene
Hazard: Flammable
Formula:
(C₆F₅)CH₂CHCH₂ MW: 208.13 g/mol Boiling Pt: 150 °C Density: 1.358 Flash Pt: 42 °C |
MDL Number:
MFCD00000301 CAS Number: 1736-60-3 UN: 1993 ADR: 3,III |
Specification Test Results
Boiling point | 150° |
Density | 1.358 |
ND20 | 1.427 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...