2-Chloro-N-[4-(trifluoromethoxy)phenyl]acetamide ≥97%

Supplier: Matrix Scientific

Synonyms: N-(Chloroacetyl)-4-(trifluoromethoxy)aniline

002929-100G 002929-25G
101776-442EA 184.62 USD
101776-442 101776-440
2-Chloro-N-[4-(trifluoromethoxy)phenyl]acetamide ≥97%
2-Chloro-N-[4-(trifluoromethoxy)phenyl]acetamide

Hazard: Irritant

Formula: C₉H₇ClF₃NO₂
MW: 253.61 g/mol
MDL Number: MFCD00219796
CAS Number: 161290-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR