Trifluoroacetyl fluoride ≥95%

Supplier: Matrix Scientific

002924-25G 002924-100G
101776-434EA 381.54 USD
101776-434 101776-436
Trifluoroacetyl fluoride ≥95%
Trifluoroacetyl fluoride

TCA listed: T
Hazard: Toxic by inhalation/Corrosive/Nonflammable Gas

Formula: C₂F₄O
MW: 116.02 g/mol
Boiling Pt: –59 °C
MDL Number: MFCD00042075
CAS Number: 354-34-7

Order Now

Specification Test Results

Boiling point -59°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR