Trifluoroacetamidine ≥85%, Technical Grade

Supplier: Matrix Scientific

002903-25G 002903-100G
101776-422EA 161.54 USD
101776-422 101776-424
Trifluoroacetamidine ≥85%, Technical Grade
Trifluoroacetamidine

Product has limited shelf life


Hazard: Corrosive/Store in Freezer/Moisture Sensitive

Formula: C₂H₃F₃N₂
Boiling Pt: 35…37 °C (11 mmHg)
Density: 1.45
MDL Number: MFCD00041545
CAS Number: 354-37-0
UN: 3267
ADR: 8,II

Order Now

Specification Test Results

Boiling point 35-37°/11m
Density 1.45
ND20 1.3801


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR