2,2,2,4'-Tetrafluoroacetophenone ≥99%
Supplier: Matrix Scientific
002872-5G
002872-25G
101776-408EA
107.69
USD
101776-408
101776-410
2,2,2,4'-Tetrafluoroacetophenone ≥99%
2,2,2,4'-Tetrafluoroacetophenone
Hazard: Flammable Solid
Formula:
C₈H₄F₄O MW: 192.1104 g/mol Boiling Pt: 66…67 °C (34 mmHg) Melting Pt: 24…27 °C Density: 1.37 |
MDL Number:
MFCD00042393 CAS Number: 655-32-3 |
Specification Test Results
Boiling point | 66-67°/34m |
Density | 1.37 |
ND20 | 1.448 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...