1H,1H,2H-Perfluoro-1-octene ≥98%

Supplier: Matrix Scientific

002862-25G 002862-100G
101776-396EA 50.77 USD
101776-396 101776-398
1H,1H,2H-Perfluoro-1-octene ≥98%
1H,1H,2H-Perfluoro-1-octene

TCA listed: T
Hazard: Flammable

Formula: C₈H₃F₁₃
Boiling Pt: 104 °C
Density: 1.52
Flash Pt: 20 °C
MDL Number: MFCD00039249
CAS Number: 25291-17-2
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Boiling point 102°
Density 1.52
ND20 1.295


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR