N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate) ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(Chloromethyl)-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane Bis(tetrafluoroborate), F-TEDA-BF4, 1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate)

002850-25G 002850-100G
101776-392EA 67.69 USD
101776-392 101776-394
N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate) ≥95%
N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate)

TCA listed: T
Hazard: Irritant/Self-heating solid/Refrigerate

Formula: C₇H₁₄B₂ClF₉N₂
MW: 354.26 g/mol
Melting Pt: 250 °C (dec.)
MDL Number: MFCD00142607
CAS Number: 140681-55-6

Order Now

Specification Test Results

Melting point 250° dec

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR