Mercury(II) fluoride ≥97%
Supplier: Matrix Scientific
Synonyms:
Mercury difluoride, Mercuric fluoride
002837-25G
002837-5G
101776-388EA
83.08
USD
101776-388
101776-386
Mercury(II) fluoride ≥97%
Mercury(II) fluoride
TCA listed: T
Hazard: Toxic/Corrosive
Formula:
F₂Hg MW: 238.59 Melting Pt: 645 Density: 8.95 |
MDL Number:
MFCD00016139 CAS Number: 7783-39-3 UN: 2025 ADR: 6.1 II Merck Index: 12,05930 |
Specification Test Results
Melting point | 645° |
Density | 8.95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...