2-Fluoro-6-iodobenzonitrile ≥98%

Supplier: Matrix Scientific

002786-5G 002786-25G
101776-378EA 15.38 USD
101776-378 101776-380
2-Fluoro-6-iodobenzonitrile ≥98%
2-Fluoro-6-iodobenzonitrile

Hazard: Toxic/Irritant-Harmful/Irritant/Light Sensitive

Formula: C₇H₃FIN
MW: 247.01 g/mol
Melting Pt: 49-51°C
MDL Number: MFCD00015478
CAS Number: 79544-29-9
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 49-51°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR