2-Chloro-6-fluorobenzoyl chloride ≥97%

Supplier: Matrix Scientific

002768-25G 002768-100G
101776-362EA 44.62 USD
101776-362 101776-364
2-Chloro-6-fluorobenzoyl chloride ≥97%
2-Chloro-6-fluorobenzoyl chloride

Hazard: Corrosive

Formula: C₇H₃Cl₂FO
MDL Number: MFCD00178746
CAS Number: 79455-63-3
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR