α,2-Dichloro-6-fluorotoluene ≥98%
Supplier: Matrix Scientific
002767-25G
002767-100G
101776-358EA
21.54
USD
101776-358
101776-360
α,2-Dichloro-6-fluorotoluene ≥98%
α,2-Dichloro-6-fluorotoluene
TCA listed: T
Hazard: Irritant/Lachrymator
Formula:
C₇H₅Cl₂F Density: 1.401 |
MDL Number:
MFCD00000896 CAS Number: 55117-15-2 UN: 3265 ADR: 8,II |
Specification Test Results
Density | 1.401 |
ND20 | 1.537 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...