2-Chloro-6-fluoroaniline ≥97%
Supplier: Matrix Scientific
002765-25G
002765-100G
101776-356EA
75.38
USD
101776-356
102550-822
2-Chloro-6-fluoroaniline ≥97%
2-Chloro-6-fluoroaniline
Hazard: Toxic
Formula:
C₆H₅ClFN MW: 145.56 g/mol Boiling Pt: 67 °C (15 mmHg) Density: 1.316 |
MDL Number:
MFCD00040309 CAS Number: 363-51-9 UN: 2810 ADR: 6.1,III |
Specification Test Results
Boiling point | 67°/15mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...