2-Chloro-6-fluoro-m-cresol

Supplier: Matrix Scientific

002761-1G 002761-5G
101776-346EA 29.23 USD
101776-346 101776-348
2-Chloro-6-fluoro-m-cresol
2-Chloro-6-fluoro-m-cresol

Hazard: Toxic/Irritant

Formula: C₇H₆ClFO
MDL Number: MFCD01631366
CAS Number: 261762-90-7
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR