2-Chloro-6-fluoro-3-methylbenzaldehyde
Supplier: Matrix Scientific
002753-5G
002753-1G
101776-336EA
106.15
USD
101776-336
101776-334
2-Chloro-6-fluoro-3-methylbenzaldehyde
2-Chloro-6-fluoro-3-methylbenzaldehyde
Hazard: Irritant/Air Sensitive
Formula:
C₈H₆ClFO Melting Pt: 26-28°C |
MDL Number:
MFCD01631360 CAS Number: 104451-99-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...