2-Fluoro-6-(trifluoromethyl)phenylacetonitrile ≥98%
Supplier: Matrix Scientific
002745-1G
002745-5G
101776-326EA
40
USD
101776-326
101776-328
2-Fluoro-6-(trifluoromethyl)phenylacetonitrile ≥98%
2-Fluoro-6-(trifluoromethyl)phenylacetonitrile
Hazard: Toxic
Formula:
C₉H₅F₄N |
MDL Number:
MFCD00061184 CAS Number: 179946-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...