2-Fluoro-6-(trifluoromethyl)benzyl bromide ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(Bromomethyl)-2-fluoro-6-(trifluoromethyl)benzene, 2-(Bromomethyl)-1-fluoro-3-(trifluoromethyl)benzene

002743-1G 002743-25G 002743-5G
101776-320EA 21.54 USD
101776-320 101776-324 101776-322
2-Fluoro-6-(trifluoromethyl)benzyl bromide ≥98%
2-Fluoro-6-(trifluoromethyl)benzyl bromide

Hazard: Corrosive/Refrigerate/Lachrymator

Formula: C₈H₅BrF₄
MW: 257.03 g/mol
Melting Pt: 38…40 °C
MDL Number: MFCD00082477
CAS Number: 239087-08-2

Order Now

Specification Test Results

Melting point 38-40°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR