2-Fluoro-6-(trifluoromethyl)benzonitrile ≥97%

Supplier: Matrix Scientific

002736-5G 002736-1G
101776-318EA 58.46 USD
101776-318 101776-316
2-Fluoro-6-(trifluoromethyl)benzonitrile ≥97%
2-Fluoro-6-(trifluoromethyl)benzonitrile

Hazard: Toxic/Irritant

Formula: C₈H₃F₄N
Melting Pt: 26…28 °C
Density: 1.373
MDL Number: MFCD00061153
CAS Number: 133116-83-3
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 26-28°
Density 1.373
ND20 1.452


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR